Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabggta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019519.2 | NP_062392.1 | 1 - 2 | 333 | 71 | |
GenBank mRNA | AF127656.1 | - | 2 - 3 | 181 | 71 | |
AF127658.1 | - | 1 - 2 | 333 | 71 | ||
AF127659.1 | - | 1 - 2 | 333 | 71 | ||
AF127660.1 | - | 2 - 3 | 181 | 71 | ||
AF127662.1 | - | 2 - 3 | 252 | 71 | ||
AK002625.1 | - | 2 - 3 | 115 | 71 | ||
AK146917.1 | - | 2 - 3 | 204 | 71 | ||
BC012214.1 | - | 2 - 3 | 186 | 3967218 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |