Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019579.3 | NP_062525.1 | 3 - 4 | 777 | 127 | |
GenBank mRNA | AF199008.1 | - | 3 - 4 | 708 | 127 | |
AK032731.1 | - | 3 - 4 | 736 | 127 | ||
AK133187.1 | - | 3 - 4 | 653 | 127 | ||
AK162014.1 | - | 3 - 4 | 736 | 127 | ||
BC067404.1 | - | 3 - 4 | 639 | 127 | ||
BC075698.1 | - | 3 - 4 | 732 | 127 | ||
BC138624.1 | - | 3 - 4 | 656 | 8861644 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |