Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000014744.mm.1 |
NM_019740.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014744.mm.1 |
NM_019740.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014744.mm.1 |
TC1000002062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014745.mm.1 |
NM_019740.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014745.mm.1 |
NM_019740.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014745.mm.1 |
TC1000002062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014755.mm.1 |
NM_019740.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014755.mm.1 |
NM_019740.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014755.mm.1 |
TC1000002062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014750.mm.1 |
NM_019740.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014750.mm.1 |
NM_019740.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014750.mm.1 |
TC1000002062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014751.mm.1 |
NM_019740.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014751.mm.1 |
NM_019740.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014751.mm.1 |
TC1000002062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014754.mm.1 |
NM_019740.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014754.mm.1 |
NM_019740.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014754.mm.1 |
TC1000002062.mm.1 |
MTA TC |