Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maged1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019791.2 | NP_062765.1 | 4 - 5 | 1628 | 77 | |
GenBank mRNA | AB029448.1 | - | 4 - 5 | 1628 | 77 | |
AF319975.1 | - | 3 - 4 | 1398 | 77 | ||
AK017275.1 | - | 4 - 5 | 1546 | 77 | ||
AK049660.1 | - | 4 - 5 | 1579 | 77 | ||
AK087937.1 | - | 4 - 5 | 1579 | 77 | ||
AK132248.1 | - | 4 - 5 | 1567 | 77 | ||
AK135452.1 | - | 4 - 5 | 1578 | 77 | ||
AK145872.1 | - | 4 - 5 | 1548 | 77 | ||
AK146867.1 | - | 4 - 5 | 1578 | 77 | ||
AK146908.1 | - | 4 - 5 | 1578 | 77 | ||
AK150876.1 | - | 1 - 2 | 567 | 77 | ||
AK195822.1 | - | 1 - 2 | 40 | 77 | ||
AK220150.1 | - | 5 - 6 | 1485 | 77 | ||
BC016438.1 | - | 4 - 5 | 1544 | 4480947 | 77 | |
BC031461.1 | - | 4 - 5 | 1568 | 4018875 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |