Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019948.2 | NP_064332.1 | 1 - 2 | 160 | 72 | |
GenBank mRNA | AB024717.1 | - | 1 - 2 | 160 | 72 | |
AK036493.1 | - | 1 - 2 | 160 | 72 | ||
AK089286.1 | - | 1 - 2 | 160 | 72 | ||
AK150528.1 | - | 1 - 2 | 160 | 72 | ||
AK150899.1 | - | 1 - 2 | 160 | 72 | ||
AK151101.1 | - | 1 - 2 | 159 | 72 | ||
AK151912.1 | - | 1 - 2 | 159 | 72 | ||
AK152333.1 | - | 1 - 2 | 160 | 72 | ||
AK153008.1 | - | 1 - 2 | 159 | 72 | ||
AK153308.1 | - | 1 - 2 | 160 | 72 | ||
AK155452.1 | - | 1 - 2 | 160 | 72 | ||
AK156437.1 | - | 1 - 2 | 160 | 72 | ||
AK170698.1 | - | 1 - 2 | 111 | 72 | ||
AK171103.1 | - | 1 - 2 | 160 | 72 | ||
BC003218.1 | - | 1 - 2 | 118 | 3158063 | 72 | |
CT010223.1 | - | 1 - 2 | 37 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |