Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rec8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020002.3 | NP_064386.2 | 16 - 17 | 1647 | 63 | |
GenBank mRNA | AF262055.1 | - | 16 - 17 | 1577 | 63 | |
AK077167.1 | - | 16 - 17 | 1582 | 63 | ||
AK146832.1 | - | 16 - 17 | 1633 | 63 | ||
BC052155.1 | - | 16 - 17 | 1643 | 6335959 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |