Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020260.2 | NP_064656.2 | 7 - 8 | 1239 | 88 | |
GenBank mRNA | AF151363.1 | - | 7 - 8 | 880 | 88 | |
AK138417.1 | - | 7 - 8 | 1479 | 88 | ||
AK149975.1 | - | 7 - 8 | 1075 | 88 | ||
AK150109.1 | - | 7 - 8 | 1075 | 88 | ||
BC138870.1 | - | 7 - 8 | 1044 | 8861892 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |