Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025578.4 | NP_079854.2 | 1 - 2 | 179 | 138 | |
GenBank mRNA | AK004037.1 | - | 1 - 2 | 158 | 138 | |
AK013196.1 | - | 1 - 2 | 139 | 138 | ||
AK131114.1 | - | 1 - 2 | 108 | 138 | ||
CB595428.1 | - | 1 - 2 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |