Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dach1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007826.3 | NP_031852.1 | 3 - 4 | 1564 | 83 | |
GenBank mRNA | AF090436.1 | - | 3 - 4 | 1108 | 83 | |
AF102547.1 | - | 3 - 4 | 1108 | 83 | ||
AJ005669.1 | - | 3 - 4 | 626 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Inventoried Probe spans exons |