Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190817.1 | NP_001177746.1 | 9 - 10 | 1424 | 83 | |
NM_007869.3 | NP_031895.1 | 9 - 10 | 1424 | 83 | ||
GenBank mRNA | AK133582.1 | - | 9 - 10 | 1406 | 83 | |
AK144228.1 | - | 9 - 10 | 1380 | 84 | ||
BC080300.1 | - | 9 - 10 | 1405 | 83 | ||
L16953.1 | - | 9 - 10 | 1101 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |