Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008130.2 | NP_032156.2 | 6 - 7 | 1254 | 124 | |
GenBank mRNA | AK144851.1 | - | 6 - 7 | 1038 | 124 | |
AK158261.1 | - | 6 - 7 | 1254 | 124 | ||
BC141135.1 | - | 6 - 7 | 955 | 9055747 | 124 | |
BC145445.1 | - | 6 - 7 | 955 | 9054003 | 124 | |
X95255.1 | - | 6 - 7 | 994 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |