Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008405.3 | NP_032431.2 | 13 - 14 | 1874 | 107 | |
GenBank mRNA | AF051367.1 | - | 13 - 14 | 1859 | 107 | |
AK036640.1 | - | 13 - 14 | 1874 | 107 | ||
AK137534.1 | - | 13 - 14 | 1875 | 107 | ||
AK158108.1 | - | 13 - 14 | 1832 | 107 | ||
BC113142.1 | - | 13 - 14 | 1843 | 40047633 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |