Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L1cam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008478.3 | NP_032504.3 | 8 - 9 | 1049 | 70 | |
GenBank mRNA | BC056988.1 | - | 8 - 9 | 974 | 6822217 | 70 |
X12875.1 | - | 7 - 8 | 796 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |