Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L1cam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008478.3 | NP_032504.3 | 21 - 22 | 2999 | 92 | |
GenBank mRNA | BC056988.1 | - | 21 - 22 | 2924 | 6822217 | 92 |
X12875.1 | - | 20 - 21 | 2746 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |