Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009331.3 | NP_033357.1 | 4 - 5 | 347 | 98 | |
GenBank mRNA | AK037616.1 | - | 4 - 5 | 348 | 98 | |
AK194540.1 | - | 1 - 2 | 99 | |||
BC056630.1 | - | 3 - 4 | 589 | 98 | ||
BC138596.1 | - | 3 - 4 | 327 | 8861616 | 98 | |
BC145343.1 | - | 3 - 4 | 327 | 9053899 | 98 | |
X61385.1 | - | 5 - 6 | 489 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |