Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB011550.1 | - | 13 - 14 | 1539 | 68 | |
AK076365.1 | - | 11 - 12 | 2455 | 68 | ||
AK133474.1 | - | 13 - 14 | 1496 | 68 | ||
AK139088.1 | - | 13 - 14 | 1745 | 68 | ||
AK144793.1 | - | 12 - 13 | 1843 | 68 | ||
BC145596.1 | - | 13 - 14 | 1417 | 68 | ||
CN456685.1 | - | 2 - 3 | 236 | 68 | ||
U81490.1 | - | 13 - 14 | 1479 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |