Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009458.4 | NP_033484.3 | 3 - 4 | 276 | 123 | |
GenBank mRNA | AK010432.1 | - | 3 - 4 | 174 | 123 | |
AK011363.1 | - | 3 - 4 | 197 | 123 | ||
AK152316.1 | - | 3 - 4 | 259 | 123 | ||
AK152645.1 | - | 3 - 4 | 259 | 123 | ||
AK153174.1 | - | 3 - 4 | 259 | 123 | ||
AK169229.1 | - | 3 - 4 | 259 | 123 | ||
BC132075.1 | - | 3 - 4 | 197 | 40130352 | 123 | |
BC132077.1 | - | 3 - 4 | 197 | 40130354 | 123 | |
U57690.1 | - | 3 - 4 | 189 | 123 | ||
X96859.1 | - | 3 - 4 | 173 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |