Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009577.3 | NP_033603.2 | 2 - 3 | 282 | 73 | |
GenBank mRNA | AK146692.1 | - | 2 - 3 | 276 | 73 | |
AK159077.1 | - | 2 - 3 | 268 | 73 | ||
BC050942.1 | - | 2 - 3 | 240 | 6821378 | 73 | |
BC067196.1 | - | 2 - 3 | 240 | 6821378 | 73 | |
BY026558.1 | - | 2 - 3 | 296 | 73 | ||
U69133.1 | - | 2 - 3 | 280 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |