Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177629.1 | NP_001171100.1 | 13 - 14 | 1467 | 67 | |
NM_010345.4 | NP_034475.2 | 14 - 15 | 1792 | 67 | ||
GenBank mRNA | AF022072.1 | - | 14 - 15 | 1766 | 67 | |
AK030727.1 | - | 14 - 15 | 1727 | 67 | ||
AK032927.1 | - | 14 - 15 | 1773 | 67 | ||
AK163841.1 | - | 14 - 15 | 1773 | 67 | ||
AK169000.1 | - | 8 - 9 | 756 | 67 | ||
BC016111.1 | - | 13 - 14 | 1467 | 4481345 | 67 | |
BC053842.1 | - | 14 - 15 | 1759 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |