Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010630.2 | NP_034760.2 | 15 - 16 | 1765 | 57 | |
GenBank mRNA | AK144304.1 | - | 16 - 17 | 1924 | 57 | |
D49545.1 | - | 15 - 16 | 1841 | 57 | ||
U92949.1 | - | 15 - 16 | 1755 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |