Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500023123.mm.1 |
TC0500002608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023123.mm.1 |
NM_001159577.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023123.mm.1 |
NM_001159577.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023123.mm.1 |
NM_001159578.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023123.mm.1 |
NM_001159578.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023123.mm.1 |
NM_001159579.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023123.mm.1 |
NM_001159579.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023123.mm.1 |
NM_001159580.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023123.mm.1 |
NM_001159580.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023123.mm.1 |
NM_010727.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023123.mm.1 |
NM_010727.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023125.mm.1 |
TC0500002608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500023125.mm.1 |
NM_001159577.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023125.mm.1 |
NM_001159577.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023125.mm.1 |
NM_001159578.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023125.mm.1 |
NM_001159578.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023125.mm.1 |
NM_001159579.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023125.mm.1 |
NM_001159579.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023125.mm.1 |
NM_001159580.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023125.mm.1 |
NM_001159580.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023125.mm.1 |
NM_010727.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500023125.mm.1 |
NM_010727.4 |
Refseq |