Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011020.3 | NP_035150.3 | 12 - 13 | 1643 | 63 | |
GenBank mRNA | AB001926.1 | - | 12 - 13 | 1609 | 63 | |
AK144225.1 | - | 12 - 13 | 1680 | 63 | ||
AK163459.1 | - | 13 - 14 | 1815 | 63 | ||
BC012712.1 | - | 12 - 13 | 1712 | 4190161 | 63 | |
BC057002.1 | - | 12 - 13 | 1643 | 5702602 | 63 | |
BC110662.1 | - | 12 - 13 | 1663 | 4511154 | 63 | |
D49482.1 | - | 12 - 13 | 1637 | 63 | ||
U23921.1 | - | 12 - 13 | 1760 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |