Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbxas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011539.3 | NP_035669.3 | 7 - 8 | 854 | 80 | |
GenBank mRNA | AK048544.1 | - | 7 - 8 | 836 | 80 | |
AK085414.1 | - | 4 - 5 | 605 | 80 | ||
AK139479.1 | - | 7 - 8 | 817 | 80 | ||
BC029014.1 | - | 7 - 8 | 856 | 5007629 | 80 | |
L18868.1 | - | 7 - 8 | 725 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |