Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tulp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011657.2 | NP_035787.1 | 5 - 6 | 622 | 76 | |
GenBank mRNA | AB016963.1 | - | 5 - 6 | 565 | 76 | |
AF045582.1 | - | 5 - 6 | 575 | 76 | ||
BC060068.1 | - | 5 - 6 | 552 | 6419643 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |