Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291804.1 | NP_001278733.1 | 10 - 11 | 1733 | 100 | |
NM_013504.5 | NP_038532.3 | 10 - 11 | 1733 | 100 | ||
GenBank mRNA | AK036893.1 | - | 10 - 11 | 1619 | 100 | |
AK079441.1 | - | 10 - 11 | 1735 | 100 | ||
AK081826.1 | - | 10 - 11 | 1641 | 100 | ||
BC109161.1 | - | 10 - 11 | 1570 | 40047883 | 100 | |
BC109162.1 | - | 10 - 11 | 1570 | 100 | ||
X97986.1 | - | 10 - 11 | 1667 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |