Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013841.3 | NP_038869.1 | 7 - 8 | 1367 | 84 | |
GenBank mRNA | AK146190.1 | - | 7 - 8 | 769 | 84 | |
AK168101.1 | - | 7 - 8 | 769 | 84 | ||
BC012691.1 | - | 7 - 8 | 743 | 4038142 | 84 | |
BC058528.1 | - | 7 - 8 | 759 | 5718352 | 84 | |
U66865.1 | - | 7 - 8 | 1382 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |