Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015756.2 | NP_056571.2 | 1 - 2 | 403 | 61 | |
GenBank mRNA | AF199421.1 | - | 1 - 2 | 403 | 61 | |
BC145113.1 | - | 1 - 2 | 213 | 9053661 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |