Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Incenp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016692.3 | NP_057901.2 | 11 - 12 | 1835 | 75 | |
GenBank mRNA | AB100432.1 | - | 10 - 11 | 1629 | 75 | |
AB100433.1 | - | 11 - 12 | 1641 | 75 | ||
AF117610.1 | - | 11 - 12 | 1771 | 75 | ||
AK034764.1 | - | 11 - 12 | 1764 | 75 | ||
AK081841.1 | - | 11 - 12 | 1769 | 75 | ||
AK088627.1 | - | 11 - 12 | 1792 | 75 | ||
BC052414.1 | - | 11 - 12 | 1768 | 6401188 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |