Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016885.2 | NP_058581.2 | 4 - 5 | 544 | 83 | |
GenBank mRNA | AF060883.1 | - | 4 - 5 | 479 | 83 | |
AK002616.1 | - | 4 - 5 | 526 | 83 | ||
AK164422.1 | - | 465 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |