Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300011656.mm.1 |
NM_001289492.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011656.mm.1 |
NM_001289493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011656.mm.1 |
NM_018734.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011656.mm.1 |
NM_018734.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011656.mm.1 |
TC0300001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011649.mm.1 |
NM_001289493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011649.mm.1 |
TC0300001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011658.mm.1 |
NM_001289492.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011658.mm.1 |
NM_001289493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011658.mm.1 |
NM_018734.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011658.mm.1 |
NM_018734.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011658.mm.1 |
TC0300001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011655.mm.1 |
NM_001289492.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011655.mm.1 |
NM_001289493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011655.mm.1 |
NM_018734.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011655.mm.1 |
NM_018734.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011655.mm.1 |
TC0300001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011662.mm.1 |
NM_001289492.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011662.mm.1 |
NM_001289493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011662.mm.1 |
NM_018734.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011662.mm.1 |
NM_018734.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011662.mm.1 |
TC0300001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011648.mm.1 |
NM_001289493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011648.mm.1 |
TC0300001446.mm.1 |
MTA TC |