Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002011.3 | NP_001002011.2 | 3 - 4 | 891 | 58 | |
NM_001111102.2 | NP_001104572.1 | 3 - 4 | 891 | 58 | ||
NM_019390.3 | NP_062263.1 | 3 - 4 | 362 | 58 | ||
GenBank mRNA | AK004619.2 | - | 3 - 4 | 846 | 58 | |
AK147150.1 | - | 3 - 4 | 847 | 58 | ||
AK149998.1 | - | 3 - 4 | 847 | 58 | ||
AK150391.1 | - | 3 - 4 | 847 | 58 | ||
AK150501.1 | - | 3 - 4 | 847 | 58 | ||
AK150624.1 | - | 3 - 4 | 847 | 58 | ||
AK152539.1 | - | 3 - 4 | 847 | 58 | ||
AK152646.1 | - | 3 - 4 | 847 | 58 | ||
AK152846.1 | - | 3 - 4 | 847 | 58 | ||
AK153295.1 | - | 3 - 4 | 789 | 58 | ||
AK161221.1 | - | 3 - 4 | 712 | 58 | ||
AK167858.1 | - | 3 - 4 | 844 | 58 | ||
BC015302.1 | - | 3 - 4 | 767 | 4240057 | 58 | |
BC094020.1 | - | 3 - 4 | 753 | 4187374 | 58 | |
D14850.1 | - | 3 - 4 | 362 | 58 | ||
DQ832702.1 | - | 3 - 4 | 642 | 58 | ||
DQ832703.1 | - | 3 - 4 | 642 | 58 | ||
X14170.1 | - | 3 - 4 | 836 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |