Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600014205.mm.1 |
NM_001252443.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014205.mm.1 |
NM_001252443.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014205.mm.1 |
NM_019678.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014205.mm.1 |
NM_019678.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014205.mm.1 |
TC1600001750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600014202.mm.1 |
NM_001252443.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014202.mm.1 |
NM_001252443.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014202.mm.1 |
NM_019678.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014202.mm.1 |
NM_019678.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014202.mm.1 |
TC1600001750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600014209.mm.1 |
NM_001252443.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014209.mm.1 |
NM_001252443.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014209.mm.1 |
NM_019678.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014209.mm.1 |
NM_019678.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014209.mm.1 |
TC1600001750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600014207.mm.1 |
NM_001252443.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014207.mm.1 |
NM_001252443.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014207.mm.1 |
NM_019678.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014207.mm.1 |
NM_019678.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014207.mm.1 |
TC1600001750.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600014208.mm.1 |
NM_001252443.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014208.mm.1 |
NM_001252443.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014208.mm.1 |
NM_019678.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014208.mm.1 |
NM_019678.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014208.mm.1 |
TC1600001750.mm.1 |
MTA TC |