Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019678.3 | NP_062652.1 | 3 - 4 | 582 | 106 | |
GenBank mRNA | AK008549.1 | - | 3 - 4 | 358 | 106 | |
AK158539.1 | - | 3 - 4 | 448 | 106 | ||
AK167475.1 | - | 3 - 4 | 446 | 106 | ||
AK186338.1 | - | 2 - 3 | 320 | 106 | ||
AK210251.1 | - | 2 - 3 | 106 | |||
BC024638.1 | - | 3 - 4 | 360 | 4015300 | 106 | |
U94662.1 | - | 3 - 4 | 484 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |