Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019793.3 | NP_062767.3 | 2 - 3 | 463 | 76 | |
GenBank mRNA | AB009021.1 | - | 2 - 3 | 245 | 76 | |
AB072450.1 | - | 2 - 3 | 273 | 76 | ||
AF133427.1 | - | 2 - 3 | 257 | 76 | ||
AF242591.1 | - | 2 - 3 | 456 | 76 | ||
AK006798.1 | - | 2 - 3 | 339 | 76 | ||
AK053325.1 | - | 2 - 3 | 314 | 76 | ||
AK089494.1 | - | 295 | 76 | |||
AK141951.1 | - | 2 - 3 | 321 | 76 | ||
AK146130.1 | - | 2 - 3 | 451 | 76 | ||
AK152175.1 | - | 2 - 3 | 348 | 76 | ||
AK154919.1 | - | 295 | 76 | |||
AK159324.1 | - | 2 - 3 | 434 | 76 | ||
AK159418.1 | - | 2 - 3 | 407 | 76 | ||
AK159621.1 | - | 2 - 3 | 418 | 76 | ||
AK168231.1 | - | 2 - 3 | 451 | 76 | ||
AK169068.1 | - | 2 - 3 | 434 | 76 | ||
BC043072.1 | - | 2 - 3 | 441 | 6404165 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |