Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aatf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019816.1 | NP_062790.1 | 11 - 12 | 1565 | 68 | |
GenBank mRNA | AF222801.1 | - | 11 - 12 | 1565 | 68 | |
AK016820.1 | - | 7 - 8 | 802 | 68 | ||
AK141334.1 | - | 11 - 12 | 1563 | 68 | ||
BC025080.1 | - | 11 - 12 | 1552 | 2648117 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |