Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ivd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019826.3 | NP_062800.1 | 2 - 3 | 255 | 67 | |
GenBank mRNA | AF225989.1 | - | 2 - 3 | 234 | 67 | |
AK005024.1 | - | 2 - 3 | 274 | 67 | ||
AK017660.1 | - | 2 - 3 | 241 | 67 | ||
AK077416.1 | - | 2 - 3 | 261 | 67 | ||
BC018325.1 | - | 2 - 3 | 245 | 4192286 | 67 | |
BC027198.1 | - | 2 - 3 | 255 | 4237982 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |