Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020046.3 | NP_064430.1 | 2 - 3 | 535 | 93 | |
GenBank mRNA | AF029667.2 | - | 2 - 3 | 240 | 93 | |
AK013102.1 | - | 2 - 3 | 487 | 93 | ||
AK088863.1 | - | 2 - 3 | 257 | 93 | ||
AK149350.1 | - | 2 - 3 | 283 | 93 | ||
BC019542.1 | - | 2 - 3 | 245 | 4039461 | 93 | |
BC027829.1 | - | 2 - 3 | 496 | 5253723 | 93 | |
BC045206.1 | - | 2 - 3 | 485 | 5253764 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |