Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmsb4x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | X16053.1 | - | 1 - 2 | 139 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Inventoried Probe spans exons |