Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021877.3 | NP_068677.2 | 17 - 18 | 3889 | 60 | |
GenBank mRNA | AB221596.1 | - | 12 - 13 | 1574 | 60 | |
AK076280.1 | - | 11 - 12 | 1374 | 60 | ||
AY312855.1 | - | 16 - 17 | 3572 | 60 | ||
AY547390.1 | - | 16 - 17 | 3569 | 60 | ||
AY547391.1 | - | 16 - 17 | 3569 | 60 | ||
BC049182.1 | - | 17 - 18 | 3889 | 6468299 | 60 | |
Z32675.1 | - | 16 - 17 | 3572 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |