Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023135.2 | NP_075624.2 | 6 - 7 | 722 | 116 | |
GenBank mRNA | AK015750.1 | - | 6 - 7 | 706 | 116 | |
BC034891.1 | - | 6 - 7 | 663 | 1479883 | 116 | |
S78182.1 | - | 6 - 7 | 705 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |