Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290437.1 | NP_001277366.1 | 4 - 5 | 1394 | 74 | |
NM_033134.3 | NP_149125.1 | 3 - 4 | 1678 | 74 | ||
NR_110957.1 | 3 - 4 | 1765 | 74 | |||
GenBank mRNA | AF226683.1 | - | 3 - 4 | 1640 | 74 | |
AK047245.1 | - | 3 - 4 | 1663 | 74 | ||
AK080075.1 | - | 3 - 4 | 1660 | 74 | ||
AK085439.1 | - | 4 - 5 | 1398 | 74 | ||
AK154097.1 | - | 3 - 4 | 1679 | 74 | ||
AK158135.1 | - | 3 - 4 | 1663 | 74 | ||
AK170174.1 | - | 4 - 5 | 1684 | 74 | ||
AK170786.1 | - | 3 - 4 | 1635 | 74 | ||
AK170812.1 | - | 3 - 4 | 1649 | 74 | ||
BC052717.1 | - | 3 - 4 | 1647 | 6837339 | 74 | |
BC057566.1 | - | 3 - 4 | 1730 | 74 | ||
BC080295.1 | - | 3 - 4 | 1643 | 30354397 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |