Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178283.4 | NP_840068.1 | 2 - 3 | 334 | 74 | |
GenBank mRNA | AF403041.1 | - | 2 - 3 | 334 | 74 | |
AK078289.1 | - | 2 - 3 | 290 | 74 | ||
AK133892.1 | - | 2 - 3 | 260 | 74 | ||
BC018240.1 | - | 2 - 3 | 288 | 4239996 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |