Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133198.2 | NP_573461.2 | 6 - 7 | 875 | 66 | |
GenBank mRNA | AF288783.1 | - | 6 - 7 | 817 | 66 | |
AK140321.1 | - | 6 - 7 | 822 | 66 | ||
AK145989.1 | - | 6 - 7 | 876 | 66 | ||
AK149491.1 | - | 1050 | 66 | |||
AK167337.1 | - | 6 - 7 | 822 | 66 | ||
BC013636.1 | - | 6 - 7 | 809 | 4196237 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |