Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007415.2 | NP_031441.2 | 15 - 16 | 2215 | 63 | |
GenBank mRNA | AK028042.1 | - | 15 - 16 | 2234 | 63 | |
AK030889.1 | - | 15 - 16 | 2218 | 63 | ||
AK088679.1 | - | 15 - 16 | 2219 | 63 | ||
AK146677.1 | - | 15 - 16 | 2236 | 63 | ||
AK159433.1 | - | 15 - 16 | 2267 | 63 | ||
AK165158.1 | - | 15 - 16 | 2235 | 63 | ||
AK166104.1 | - | 15 - 16 | 2267 | 63 | ||
AK167699.1 | - | 15 - 16 | 2207 | 63 | ||
AK169326.1 | - | 15 - 16 | 2236 | 63 | ||
BC012041.1 | - | 15 - 16 | 2215 | 2648390 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |