Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008239.4 | NP_032265.3 | 1 | 1500 | 138 | |
GenBank mRNA | AK147202.1 | - | 1 | 1506 | 138 | |
BC047155.1 | - | 1 | 1233 | 6489152 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon |