Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008485.3 | NP_032511.3 | 18 - 19 | 2912 | 79 | |
GenBank mRNA | U43327.2 | - | 17 - 18 | 2642 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |