Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008707.3 | NP_032733.1 | 1 - 2 | 141 | 123 | |
GenBank mRNA | AF043326.1 | - | 1 - 2 | 153 | 123 | |
AK146523.1 | - | 1 - 2 | 156 | 123 | ||
BC016526.1 | - | 1 - 2 | 141 | 3589678 | 123 | |
BC021635.1 | - | 1 - 2 | 155 | 3989540 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |