Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011715.2 | NP_035845.1 | 10 - 11 | 1260 | 72 | |
GenBank mRNA | AF020055.1 | - | 10 - 11 | 1260 | 72 | |
AK004644.1 | - | 10 - 11 | 1368 | 72 | ||
AK004858.1 | - | 10 - 11 | 1331 | 72 | ||
AK151146.1 | - | 10 - 11 | 1257 | 72 | ||
AK153338.1 | - | 10 - 11 | 1271 | 72 | ||
AK156300.1 | - | 10 - 11 | 1297 | 72 | ||
AK165225.1 | - | 10 - 11 | 1230 | 72 | ||
AK165323.1 | - | 10 - 11 | 1231 | 72 | ||
AK167241.1 | - | 10 - 11 | 1286 | 72 | ||
AK167275.1 | - | 10 - 11 | 1328 | 72 | ||
AK167813.1 | - | 10 - 11 | 1258 | 72 | ||
AK169835.1 | - | 10 - 11 | 1354 | 72 | ||
AK169893.1 | - | 10 - 11 | 1290 | 72 | ||
AK170379.1 | - | 10 - 11 | 1270 | 72 | ||
AK177180.1 | - | 2 - 3 | 257 | 72 | ||
AK206614.1 | - | 1 - 2 | 32 | 72 | ||
BC049117.1 | - | 10 - 11 | 1352 | 72 | ||
BC119358.1 | - | 10 - 11 | 1216 | 8734107 | 72 | |
BC125295.1 | - | 10 - 11 | 1216 | 40129810 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |