Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827192 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827193 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827190 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827191 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827188 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827189 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1975011 |
AK003070.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1975011 |
AK003897.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1975011 |
NM_025388.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1975011 |
BC021936.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1975011 |
BC055109.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
426258 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
594937 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760958 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
281259 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
649639 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
888313 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
426632 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
956706 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
618862 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
218057 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882692 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
124966 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
344972 |
AK003544.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
344972 |
AK147538.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
344972 |
AK154627.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
344972 |
BC003293.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
344972 |
BC007481.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
344972 |
BC036274.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
344972 |
XM_001003435.2 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
344972 |
XM_912589.3 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
545262 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
71287 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
404988 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
144193 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
665191 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
782744 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
560048 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
159953 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
687513 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
415210 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
62462 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
698893 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
229000 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
313359 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
546565 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
951748 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
71854 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385832 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
58794 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
266082 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
162726 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
235414 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
736568 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
771801 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
166501 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
151202 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
109943 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
98428 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
178432 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
63862 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P124605 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P114606 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1975011 |
AK003070.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1975011 |
AK003897.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1975011 |
NM_025388.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1975011 |
BC021936.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1975011 |
BC055109.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827184 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827165 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827177 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827159 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827199 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827194 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827170 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827172 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827197 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827195 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827166 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827164 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827190 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827169 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827183 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827174 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827208 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827181 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827187 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827202 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827158 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827193 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827196 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827203 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827180 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827189 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827205 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827210 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20827162 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04354051 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00250721 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04354377 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04354061 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04354205 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00250939 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00250944 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00250818 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P124605 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04354404 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P114606 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04354079 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04354067 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00250731 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P512744 |
Y17677.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P512744 |
AK003070.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P512744 |
AK003897.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P512744 |
NM_025388.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P512744 |
AK011260.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P123301 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P00250939 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04354051 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04354079 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04354205 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P124605 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P114606 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00289281 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01092279 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01092285 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01117298 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01092274 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827192 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827190 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827188 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827189 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827191 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20827193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827162 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827120 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827079 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827194 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827187 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827164 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827097 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827166 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827156 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827180 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827169 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827213 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827119 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827189 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827145 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827093 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827142 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827074 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827197 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827202 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827112 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827127 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827077 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827129 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827086 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827170 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827135 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827203 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827158 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827174 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827094 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20827083 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827158 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827166 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01117298 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827224 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01092280 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827232 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827170 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00353167 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00489113 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827229 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827231 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00289250 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827172 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827162 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827196 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827189 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00353166 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827205 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827206 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827190 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827174 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00293673 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827227 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00825212 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827187 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00289285 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827169 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827193 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827210 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827226 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00293678 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827202 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01141829 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827197 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827203 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827222 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00014027 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827230 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01092288 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827181 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827213 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20827184 |
- |
- |