Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290706.1 | NP_001277635.1 | 3 - 4 | 439 | 70 | |
NM_025452.4 | NP_079728.2 | 3 - 4 | 457 | 70 | ||
GenBank mRNA | AK008935.1 | - | 3 - 4 | 372 | 70 | |
BC019563.1 | - | 3 - 4 | 386 | 4219997 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |